siRNA Details
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008749.2 | NP_001008749.2 | 2 | 342 | ||
XM_005249996.3 | XP_005250053.1 | 2 | 625 | |||
XM_005249997.3 | XP_005250054.1 | 2 | 644 | |||
XM_005249998.3 | XP_005250055.1 | 2 | 645 | |||
XM_017012207.1 | XP_016867696.1 | 2 | 538 | |||
GenBank mRNA | AF091033.1 | - | 1 | 60 | ||
BC140796.1 | - | 1 | 212 | 9021664 | ||
BC171745.1 | - | 1 | 212 | |||
DN996121.1 | - | 2 | 316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
