siRNA Details
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008749.2 | NP_001008749.2 | 2 | 365 | ||
XM_005249996.3 | XP_005250053.1 | 2 | 648 | |||
XM_017012207.1 | XP_016867696.1 | 2 | 561 | |||
GenBank mRNA | AF091033.1 | - | 1 | 83 | ||
BC140796.1 | - | 1 | 235 | 9021664 | ||
BC171745.1 | - | 1 | 235 | |||
DN996121.1 | - | 2 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
