siRNA Details
Gene Symbol: ASPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020164.4 | NP_064549.1 | 6 | 798 | ||
NM_032467.3 | NP_115856.1 | 5 | 753 | |||
GenBank mRNA | AF184241.1 | - | 6 | 678 | ||
AF224468.1 | - | 5 | 703 | |||
AF224469.1 | - | 6 | 723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
