siRNA Details
Gene Symbol: WSCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015253.1 | NP_056068.1 | 9 | 1721 | ||
XM_005256572.1 | XP_005256629.1 | 9 | 1809 | |||
XM_005256573.1 | XP_005256630.1 | 9 | 1784 | |||
GenBank mRNA | AB011095.1 | - | 8 | 1073 | ||
AK000243.1 | - | 7 | 1194 | |||
AK289603.1 | - | 9 | 1780 | |||
AK309645.1 | - | 8 | 1173 | |||
AL833730.1 | - | 9 | 1721 | |||
BC009975.2 | - | 9 | 1746 | 4110708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
