siRNA Details
Gene Symbol: WSCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015253.1 | NP_056068.1 | 2 | 689 | ||
XM_005256572.1 | XP_005256629.1 | 2 | 777 | |||
XM_005256573.1 | XP_005256630.1 | 2 | 752 | |||
XM_011523777.1 | XP_011522079.1 | 2 | 777 | |||
GenBank mRNA | AB011095.1 | - | 1 | 41 | ||
AK000243.1 | - | 1 | 277 | |||
AK289603.1 | - | 2 | 748 | |||
AL833730.1 | - | 2 | 689 | |||
BC009975.2 | - | 2 | 714 | 4110708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
