siRNA Details
Gene Symbol: WSCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015253.1 | NP_056068.1 | 6 | 1311 | ||
XM_005256572.1 | XP_005256629.1 | 6 | 1399 | |||
XM_005256573.1 | XP_005256630.1 | 6 | 1374 | |||
GenBank mRNA | AB011095.1 | - | 5 | 663 | ||
AK000243.1 | - | 4 | 784 | |||
AK289603.1 | - | 6 | 1370 | |||
AK309645.1 | - | 5 | 763 | |||
AL833730.1 | - | 6 | 1311 | |||
BC009975.2 | - | 6 | 1336 | 4110708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
