siRNA Details
Gene Symbol: SON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291411.1 | NP_001278340.1 | 3 | 3417 | ||
NM_032195.2 | NP_115571.2 | 3 | 3417 | |||
NM_138927.2 | NP_620305.2 | 3 | 3417 | |||
NR_103797.1 | 3 | 3417 | ||||
GenBank mRNA | AB028942.2 | - | 3 | 3386 | ||
AF380179.1 | - | 4 | 2434 | |||
AF380180.1 | - | 3 | 3391 | |||
AF380181.1 | - | 3 | 3391 | |||
AF380183.1 | - | 3 | 3391 | |||
AF380184.1 | - | 3 | 3391 | |||
AK074269.1 | - | 1 | 2379 | |||
AK128071.1 | - | 1 | 2364 | |||
AK307612.1 | - | 3 | 3417 | |||
AY026895.1 | - | 4 | 3291 | |||
BC002422.2 | - | 1 | 1881 | |||
X63751.1 | - | 1 | 1358 | |||
X63753.1 | - | 2 | 752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
