siRNA Details
Gene Symbol: DSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001941.4 | NP_001932.2 | 10 | 1628 | ||
NM_024423.3 | NP_077741.2 | 10 | 1628 | |||
GenBank mRNA | AK291748.1 | - | 10 | 1554 | ||
D17427.1 | - | 10 | 1540 | |||
X83929.1 | - | 11 | 1494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
