siRNA Details
Gene Symbol: PDLIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005451.4 | NP_005442.2 | 3 | 241 | ||
NM_203352.2 | NP_976227.1 | 3 | 241 | |||
NM_213636.2 | NP_998801.1 | 3 | 241 | |||
NR_103804.1 | 3 | 241 | ||||
XM_011534698.2 | XP_011533000.1 | 3 | 237 | |||
XM_011534699.2 | XP_011533001.1 | 3 | 209 | |||
GenBank mRNA | AF265209.1 | - | 3 | 236 | ||
AF345904.1 | - | 3 | 197 | |||
AF345905.1 | - | 3 | 197 | |||
AF345906.1 | - | 3 | 197 | |||
BC001093.2 | - | 3 | 219 | 3506748 | ||
BC014521.2 | - | 3 | 209 | |||
BC067806.1 | - | 3 | 237 | |||
BC084575.1 | - | 3 | 219 | 6729368 | ||
BG714861.1 | - | 3 | 242 | |||
BG772677.1 | - | 3 | 245 | |||
CB993539.1 | - | 267 | ||||
HY026171.1 | - | 3 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
