siRNA Details
Gene Symbol: SIAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006610.1 | NP_001006611.1 | 2 | 1423 | ||
NM_003031.3 | NP_003022.3 | 2 | 1051 | |||
XM_006721246.1 | XP_006721309.1 | 3 | 1227 | |||
XM_011523279.1 | XP_011521581.1 | 6 | 2567 | |||
GenBank mRNA | AK094663.1 | - | 1 | 2388 | ||
AK225895.1 | - | 2 | 1084 | |||
BC035562.1 | - | 2 | 1051 | 5224755 | ||
BC042550.1 | - | 2 | 1393 | 5198176 | ||
BX647064.1 | - | 1 | 1995 | |||
U63295.1 | - | 1 | 934 | |||
U76247.1 | - | 2 | 1120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
