siRNA Details
Gene Symbol: SIAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006610.1 | NP_001006611.1 | 2 | 1567 | ||
NM_003031.3 | NP_003022.3 | 2 | 1195 | |||
XM_006721246.1 | XP_006721309.1 | 3 | 1371 | |||
XM_011523279.1 | XP_011521581.1 | 6 | 2711 | |||
GenBank mRNA | AK094663.1 | - | 1 | 2532 | ||
AK225895.1 | - | 2 | 1228 | |||
BC035562.1 | - | 2 | 1195 | 5224755 | ||
BC042550.1 | - | 2 | 1537 | 5198176 | ||
BX647064.1 | - | 1 | 2139 | |||
U63295.1 | - | 1 | 1078 | |||
U76247.1 | - | 2 | 1264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
