siRNA Details
Gene Symbol: SMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003652.3 | NP_001003652.1 | 8 | 1212 | ||
NM_001135937.2 | NP_001129409.1 | 7 | 1122 | |||
NM_005901.5 | NP_005892.1 | 8 | 1104 | |||
XM_005258259.3 | XP_005258316.1 | 9 | 1345 | |||
XM_006722451.3 | XP_006722514.1 | 9 | 1357 | |||
XM_011525984.2 | XP_011524286.1 | 8 | 701 | |||
XM_011525985.2 | XP_011524287.1 | 10 | 1404 | |||
XM_017025745.1 | XP_016881234.1 | 10 | 1443 | |||
XM_017025746.1 | XP_016881235.1 | 8 | 1255 | |||
XM_017025747.1 | XP_016881236.1 | 9 | 1330 | |||
XM_017025748.1 | XP_016881237.1 | 10 | 1416 | |||
XM_017025750.1 | XP_016881239.1 | 9 | 1305 | |||
GenBank mRNA | AF027964.1 | - | 8 | 918 | ||
AK299218.1 | - | 7 | 818 | |||
AK312963.1 | - | 7 | 941 | |||
BC014840.1 | - | 8 | 1060 | 4449540 | ||
BC025699.1 | - | 8 | 918 | 5221801 | ||
BT007422.1 | - | 7 | 812 | |||
U59911.1 | - | 8 | 911 | |||
U65019.1 | - | 7 | 848 | |||
U68018.1 | - | 7 | 867 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
