siRNA Details
Gene Symbol: UHRF1BP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006947.1 | NP_001006948.1 | 12 | 1725 | ||
NM_015054.1 | NP_055869.1 | 12 | 1725 | |||
XM_005268738.2 | XP_005268795.1 | 13 | 1831 | |||
XM_005268739.3 | XP_005268796.1 | 11 | 1502 | |||
XM_011538054.2 | XP_011536356.1 | 2 | 200 | |||
XM_017019047.1 | XP_016874536.1 | 12 | 1685 | |||
XM_017019048.1 | XP_016874537.1 | 12 | 1648 | |||
XM_017019049.1 | XP_016874538.1 | 12 | 1685 | |||
XM_017019050.1 | XP_016874539.1 | 12 | 1630 | |||
XM_017019051.1 | XP_016874540.1 | 13 | 1776 | |||
GenBank mRNA | AB014601.2 | - | 8 | 1166 | ||
AK026472.1 | - | 3 | 322 | |||
AK125813.1 | - | 12 | 1685 | |||
AL136821.1 | - | 5 | 530 | |||
AL833850.1 | - | 2 | 179 | |||
BC014891.1 | - | 12 | 1708 | |||
BC026286.1 | - | 5 | 492 | |||
BC127017.1 | - | 12 | 1543 | 40124660 | ||
BX647380.1 | - | 6 | 696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
