siRNA Details
Gene Symbol: BAGE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC107039.2 | - | 2 | 200 | ||
BC113810.1 | - | 2 | 200 |
Gene Symbol: BAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182482.2 | NP_872288.2 | 2 | 305 | ||
GenBank mRNA | AF218570.1 | - | 2 | 305 | ||
AK292540.1 | - | 2 | 345 | |||
BC101132.2 | - | 2 | 200 | |||
BC101133.2 | - | 2 | 200 |
Gene Symbol: BAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182481.1 | NP_872287.1 | 2 | 305 | ||
GenBank mRNA | AF339514.1 | - | 2 | 305 | ||
BC101134.2 | - | 2 | 200 | 40014666 |
Gene Symbol: LINC01189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046203.2 | 2 | 539 | |||
GenBank mRNA | BC032596.1 | - | 2 | 550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
