siRNA Details
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018014.3 | NP_060484.2 | 4 | 2236 | ||
NM_022893.3 | NP_075044.2 | 4 | 2236 | |||
XM_011532909.1 | XP_011531211.1 | 4 | 2236 | |||
XM_011532910.1 | XP_011531212.1 | 4 | 2236 | |||
XM_011532912.1 | XP_011531214.1 | 3 | 2134 | |||
XM_017004333.1 | XP_016859822.1 | 4 | 2004 | |||
XM_017004334.1 | XP_016859823.1 | 3 | 2134 | |||
XM_017004335.1 | XP_016859824.1 | 3 | 1944 | |||
XM_017004336.1 | XP_016859825.1 | 4 | 1799 | |||
XM_017004337.1 | XP_016859826.1 | 3 | 8705 | |||
XM_017004338.1 | XP_016859827.1 | 3 | 4449 | |||
GenBank mRNA | AB058712.1 | - | 4 | 2136 | ||
AF080216.1 | - | 6 | 2316 | |||
AJ404611.1 | - | 4 | 2236 | |||
AJ404612.1 | - | 4 | 2236 | |||
AK001035.1 | - | 1 | 105 | |||
AK055251.1 | - | 4 | 2236 | |||
AK092316.1 | - | 3 | 1675 | |||
AK297507.1 | - | 3 | 2046 | |||
AK300012.1 | - | 6 | 1927 | |||
AY228763.1 | - | 3 | 2067 | |||
BC021098.1 | - | 4 | 2117 | 5087967 | ||
JN852960.1 | - | 1 | 658 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
