siRNA Details
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018014.3 | NP_060484.2 | 4 | 2210 | ||
NM_022893.3 | NP_075044.2 | 4 | 2210 | |||
XM_011532909.1 | XP_011531211.1 | 4 | 2210 | |||
XM_011532910.1 | XP_011531212.1 | 4 | 2210 | |||
XM_011532912.1 | XP_011531214.1 | 3 | 2108 | |||
XM_017004333.1 | XP_016859822.1 | 4 | 1978 | |||
XM_017004334.1 | XP_016859823.1 | 3 | 2108 | |||
XM_017004335.1 | XP_016859824.1 | 3 | 1918 | |||
XM_017004336.1 | XP_016859825.1 | 4 | 1773 | |||
XM_017004337.1 | XP_016859826.1 | 3 | 8679 | |||
XM_017004338.1 | XP_016859827.1 | 3 | 4423 | |||
GenBank mRNA | AB058712.1 | - | 4 | 2110 | ||
AF080216.1 | - | 6 | 2290 | |||
AJ404611.1 | - | 4 | 2210 | |||
AJ404612.1 | - | 4 | 2210 | |||
AK001035.1 | - | 1 | 79 | |||
AK055251.1 | - | 4 | 2210 | |||
AK092316.1 | - | 3 | 1649 | |||
AK297507.1 | - | 3 | 2020 | |||
AK300012.1 | - | 6 | 1901 | |||
AY228763.1 | - | 3 | 2041 | |||
BC021098.1 | - | 4 | 2091 | 5087967 | ||
JN852960.1 | - | 1 | 632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
