siRNA Details
Gene Symbol: INTS6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182540.4 | NP_872346.3 | 17 | 2854 | ||
XM_005262394.2 | XP_005262451.1 | 18 | 2965 | |||
XM_006724740.2 | XP_006724803.1 | 18 | 2965 | |||
XM_006724741.3 | XP_006724804.1 | 18 | 2833 | |||
XM_017029342.1 | XP_016884831.1 | 17 | 2854 | |||
XM_017029343.1 | XP_016884832.1 | 17 | 2722 | |||
XM_017029344.1 | XP_016884833.1 | 17 | 2722 | |||
XM_017029345.1 | XP_016884834.1 | 17 | 2651 | |||
GenBank mRNA | AK096544.1 | - | 16 | 2560 | ||
AK123209.1 | - | 6 | 878 | |||
AK126202.1 | - | 3 | 2783 | |||
AL833319.1 | - | 17 | 5824 | |||
BC140715.1 | - | 17 | 2676 | 9021583 | ||
BX648113.1 | - | 16 | 6113 | |||
CR936620.1 | - | 16 | 6066 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
