siRNA Details
Gene Symbol: INTS6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182540.4 | NP_872346.3 | 16 | 2806 | ||
XM_005262394.2 | XP_005262451.1 | 17 | 2917 | |||
XM_006724740.2 | XP_006724803.1 | 17 | 2917 | |||
XM_006724741.3 | XP_006724804.1 | 17 | 2785 | |||
XM_017029342.1 | XP_016884831.1 | 16 | 2806 | |||
XM_017029343.1 | XP_016884832.1 | 16 | 2674 | |||
XM_017029344.1 | XP_016884833.1 | 16 | 2674 | |||
XM_017029345.1 | XP_016884834.1 | 16 | 2603 | |||
GenBank mRNA | AK096544.1 | - | 15 | 2512 | ||
AK123209.1 | - | 5 | 830 | |||
AK126202.1 | - | 2 | 2735 | |||
BC140715.1 | - | 16 | 2628 | 9021583 | ||
BX648113.1 | - | 15 | 6065 | |||
CR936620.1 | - | 15 | 6018 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
