siRNA Details
Gene Symbol: C4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252204.1 | NP_001239133.1 | 33 | 4295 | ||
NM_007293.2 | NP_009224.2 | 33 | 4295 | |||
GenBank mRNA | AB209989.1 | - | 33 | 4284 | ||
AK293191.1 | - | 8 | 940 | |||
BC063289.1 | - | 33 | 4328 | 5289068 | ||
BC144546.1 | - | 33 | 4265 | 9053073 | ||
BC146673.1 | - | 33 | 4284 | 40148477 | ||
BC146849.1 | - | 33 | 4265 | 9056777 | ||
BC151204.1 | - | 33 | 4282 | 40147083 | ||
BC171786.1 | - | 33 | 4265 | 9054440 | ||
K02403.1 | - | 33 | 4284 |
Gene Symbol: C4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002029.3 | NP_001002029.3 | 33 | 4295 | ||
GenBank mRNA | AK304564.1 | - | 12 | 1410 |
Gene Symbol: C4B_2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242823.2 | NP_001229752.1 | 33 | 4295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
