siRNA Details
Gene Symbol: GYPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256584.1 | NP_001243513.1 | 5 | 1549 | ||
NM_002101.4 | NP_002092.1 | 4 | 731 | |||
NM_016815.3 | NP_058131.1 | 3 | 674 | |||
XM_006712460.3 | XP_006712523.1 | 4 | 599 | |||
GenBank mRNA | BC104246.1 | - | 4 | 532 | 40033899 | |
BC104247.1 | - | 4 | 532 | 40033903 | ||
BC106051.1 | - | 4 | 563 | 4707035 | ||
CB992018.1 | - | 3 | 459 | |||
M28335.1 | - | 4 | 396 | |||
M36284.1 | - | 4 | 402 | |||
X12496.1 | - | 4 | 522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
