siRNA Details
Gene Symbol: THADA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083953.1 | NP_001077422.1 | 30 | 4658 | ||
NM_022065.4 | NP_071348.3 | 30 | 4446 | |||
NR_073394.1 | 28 | 4256 | ||||
XM_006712061.3 | XP_006712124.1 | 31 | 4493 | |||
XM_006712062.1 | XP_006712125.1 | 30 | 4443 | |||
XM_006712063.1 | XP_006712126.1 | 30 | 4326 | |||
XM_006712064.2 | XP_006712127.1 | 30 | 4446 | |||
XM_006712065.1 | XP_006712128.1 | 28 | 4224 | |||
XM_006712066.1 | XP_006712129.1 | 28 | 4083 | |||
XM_006712067.1 | XP_006712130.1 | 28 | 4080 | |||
XM_006712068.3 | XP_006712131.1 | 30 | 4446 | |||
XM_006712069.3 | XP_006712132.1 | 15 | 1954 | |||
XM_017004674.1 | XP_016860163.1 | 30 | 4323 | |||
XM_017004675.1 | XP_016860164.1 | 29 | 4198 | |||
GenBank mRNA | AB051554.2 | - | 30 | 4658 | ||
AF323176.1 | - | 3 | 376 | |||
AK025445.1 | - | 15 | 2025 | |||
AK025530.1 | - | 5 | 519 | |||
AK126824.1 | - | 18 | 2712 | |||
AK290023.1 | - | 30 | 4446 | |||
AL832141.1 | - | 30 | 4428 | |||
AY149629.1 | - | 30 | 4438 | |||
AY149633.1 | - | 28 | 4248 | |||
BC007743.1 | - | 1 | 40 | |||
BC025773.1 | - | 6 | 659 | |||
BC105976.1 | - | 9 | 956 | |||
BX648155.1 | - | 30 | 4442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
