siRNA Details
Gene Symbol: THADA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083953.1 | NP_001077422.1 | 38 | 6065 | ||
NM_022065.4 | NP_071348.3 | 38 | 5853 | |||
NR_073394.1 | 36 | 5663 | ||||
XM_006712061.3 | XP_006712124.1 | 39 | 5900 | |||
XM_006712062.1 | XP_006712125.1 | 38 | 5850 | |||
XM_006712063.1 | XP_006712126.1 | 38 | 5733 | |||
XM_006712064.2 | XP_006712127.1 | 37 | 5721 | |||
XM_006712065.1 | XP_006712128.1 | 36 | 5631 | |||
XM_006712066.1 | XP_006712129.1 | 36 | 5490 | |||
XM_006712067.1 | XP_006712130.1 | 36 | 5487 | |||
XM_006712069.3 | XP_006712132.1 | 23 | 3361 | |||
XM_017004674.1 | XP_016860163.1 | 38 | 5730 | |||
XM_017004675.1 | XP_016860164.1 | 37 | 5605 | |||
GenBank mRNA | AB051554.2 | - | 38 | 6065 | ||
AF323176.1 | - | 11 | 1783 | |||
AK025445.1 | - | 23 | 3432 | |||
AK025530.1 | - | 13 | 1926 | |||
AK290023.1 | - | 38 | 5853 | |||
AY149629.1 | - | 38 | 5845 | |||
AY149633.1 | - | 36 | 5655 | |||
BC007743.1 | - | 9 | 1447 | |||
BC025773.1 | - | 14 | 2066 | |||
BC105976.1 | - | 17 | 2363 | |||
BX648155.1 | - | 38 | 5849 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
