siRNA Details
Gene Symbol: THADA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083953.1 | NP_001077422.1 | 31 | 4760 | ||
NM_022065.4 | NP_071348.3 | 31 | 4548 | |||
NR_073394.1 | 29 | 4358 | ||||
XM_006712061.3 | XP_006712124.1 | 32 | 4595 | |||
XM_006712062.1 | XP_006712125.1 | 31 | 4545 | |||
XM_006712063.1 | XP_006712126.1 | 31 | 4428 | |||
XM_006712064.2 | XP_006712127.1 | 31 | 4548 | |||
XM_006712065.1 | XP_006712128.1 | 29 | 4326 | |||
XM_006712066.1 | XP_006712129.1 | 29 | 4185 | |||
XM_006712067.1 | XP_006712130.1 | 29 | 4182 | |||
XM_006712069.3 | XP_006712132.1 | 16 | 2056 | |||
XM_017004674.1 | XP_016860163.1 | 31 | 4425 | |||
XM_017004675.1 | XP_016860164.1 | 30 | 4300 | |||
GenBank mRNA | AB051554.2 | - | 31 | 4760 | ||
AF323176.1 | - | 4 | 478 | |||
AK025445.1 | - | 16 | 2127 | |||
AK025530.1 | - | 6 | 621 | |||
AK290023.1 | - | 31 | 4548 | |||
AY149629.1 | - | 31 | 4540 | |||
AY149633.1 | - | 29 | 4350 | |||
BC007743.1 | - | 2 | 142 | |||
BC025773.1 | - | 7 | 761 | |||
BC105976.1 | - | 10 | 1058 | |||
BX648155.1 | - | 31 | 4544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
