siRNA Details
Gene Symbol: ZNF727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159522.1 | NP_001152994.1 | 2 | 200 | ||
XM_017012225.1 | XP_016867714.1 | 2 | 837 | |||
GenBank mRNA | DA235663.1 | - | 2 | 200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
