siRNA Details
Gene Symbol: LOC100499484-C9ORF174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036527.1 | 33 | 4013 | |||
NR_036528.1 | 33 | 4013 | ||||
NR_036529.1 | 30 | 3573 |
Gene Symbol: CCDC180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020893.2 | NP_065944.2 | 19 | 2718 | ||
GenBank mRNA | AK302965.1 | - | 20 | 2798 | ||
BC047097.1 | - | 20 | 2661 | |||
CR749320.1 | - | 19 | 2660 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
