siRNA Details
Gene Symbol: CCDC180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020893.2 | NP_065944.2 | 37 | 5555 | ||
GenBank mRNA | CR627453.1 | - | 3 | 6575 |
Gene Symbol: LOC100499484-C9ORF174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036527.1 | 49 | 6268 | |||
NR_036528.1 | 51 | 6850 | ||||
NR_036529.1 | 45 | 5717 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
