siRNA Details
Gene Symbol: NUP210P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034158.1 | 4, 5 | 648 | |||
GenBank mRNA | BC042038.1 | - | 4, 5 | 648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
