siRNA Details
Gene Symbol: UBXN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077262.1 | NP_001070730.1 | 3 | 238 | ||
NM_145345.2 | NP_663320.2 | 4 | 457 | |||
NM_183008.2 | NP_892120.2 | 4 | 363 | |||
GenBank mRNA | AF520989.1 | - | 3 | 166 | ||
AF521017.1 | - | 3 | 167 | |||
AK093258.1 | - | 4 | 307 | |||
AK094198.1 | - | 3 | 170 | |||
AK097368.1 | - | 4 | 555 | |||
AK291503.1 | - | 3 | 162 | |||
AK296179.1 | - | 2 | 64 | |||
AY007163.1 | - | 2 | 48 | |||
BC038106.1 | - | 3 | 175 | 5269682 | ||
BX647774.1 | - | 3 | 171 | |||
BX648412.1 | - | 3 | 171 | |||
DA421971.1 | - | 3 | 170 | |||
DA989799.1 | - | 3 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
