siRNA Details
Gene Symbol: UBXN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077262.1 | NP_001070730.1 | 7 | 657 | ||
NM_145345.2 | NP_663320.2 | 11 | 1137 | |||
NM_183008.2 | NP_892120.2 | 12 | 1142 | |||
GenBank mRNA | AF370425.1 | - | 1 | 931 | ||
AF520989.1 | - | 10 | 846 | |||
AF521017.1 | - | 10 | 847 | |||
AF521018.1 | - | 10 | 834 | |||
AK074358.1 | - | 6 | 1615 | |||
AK090792.1 | - | 6 | 1616 | |||
AK093258.1 | - | 12 | 1086 | |||
AK094198.1 | - | 7 | 589 | |||
AK097368.1 | - | 11 | 1235 | |||
AY007163.1 | - | 8 | 601 | |||
BC038106.1 | - | 10 | 1935 | 5269682 | ||
BX647774.1 | - | 10 | 1931 | |||
BX648412.1 | - | 9 | 1832 | |||
CR749413.1 | - | 10 | 1929 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
