siRNA Details
Gene Symbol: UBXN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077262.1 | NP_001070730.1 | 2 | 182 | ||
NM_145345.2 | NP_663320.2 | 3 | 401 | |||
NM_183008.2 | NP_892120.2 | 3 | 307 | |||
GenBank mRNA | AF520989.1 | - | 2 | 110 | ||
AF521017.1 | - | 2 | 111 | |||
AF521018.1 | - | 2 | 99 | |||
AK093258.1 | - | 3 | 251 | |||
AK094198.1 | - | 2 | 114 | |||
AK097368.1 | - | 3 | 499 | |||
AK291503.1 | - | 2 | 106 | |||
BC038106.1 | - | 2 | 119 | 5269682 | ||
BX647774.1 | - | 2 | 115 | |||
BX648412.1 | - | 2 | 115 | |||
CR749413.1 | - | 2 | 113 | |||
DA421971.1 | - | 2 | 114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
