siRNA Details
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000899.4 | NP_000890.1 | 10 | 1070 | ||
NM_003994.5 | NP_003985.2 | 9 | 986 | |||
GenBank mRNA | AF400436.1 | - | 10 | 1059 | ||
AK290319.1 | - | 9 | 982 | |||
AK293002.1 | - | 9 | 902 | |||
BC069733.1 | - | 10 | 1031 | 7262647 | ||
BC069783.1 | - | 10 | 1032 | 7262659 | ||
BC069797.1 | - | 10 | 1027 | 7262576 | ||
BC126166.1 | - | 10 | 1025 | 8991882 | ||
BC143899.1 | - | 9 | 941 | 9052417 | ||
CR749222.1 | - | 9 | 818 | |||
M59964.1 | - | 10 | 1059 | |||
S45995.1 | - | 2 | 109 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013598, S40536, X99322, M64262, U44725, and others.
Gene Ontology Categories:
