siRNA Details
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000899.4 | NP_000890.1 | 7 | 883 | ||
NM_003994.5 | NP_003985.2 | 6 | 799 | |||
GenBank mRNA | AF119835.1 | - | 6 | 746 | ||
AF400436.1 | - | 7 | 871 | |||
AF400437.1 | - | 6 | 746 | |||
AK290319.1 | - | 6 | 795 | |||
AK293002.1 | - | 6 | 715 | |||
BC069733.1 | - | 7 | 844 | 7262647 | ||
BC069783.1 | - | 7 | 845 | 7262659 | ||
BC069797.1 | - | 7 | 840 | 7262576 | ||
BC074725.2 | - | 7 | 728 | 30915176 | ||
BC126166.1 | - | 7 | 838 | 8991882 | ||
BC143899.1 | - | 6 | 754 | 9052417 | ||
CR749222.1 | - | 6 | 631 | |||
M59964.1 | - | 7 | 871 | |||
S42571.1 | - | 2 | 106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
