siRNA Details
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000899.4 | NP_000890.1 | 9 | 1008 | ||
NM_003994.5 | NP_003985.2 | 8 | 924 | |||
GenBank mRNA | AK290319.1 | - | 8 | 920 | ||
AK293002.1 | - | 8 | 840 | |||
BC069733.1 | - | 9 | 969 | 7262647 | ||
BC069783.1 | - | 9 | 970 | 7262659 | ||
BC069797.1 | - | 9 | 965 | 7262576 | ||
BC074725.2 | - | 9 | 853 | 30915176 | ||
BC126166.1 | - | 9 | 963 | 8991882 | ||
BC143899.1 | - | 8 | 879 | 9052417 | ||
CR749222.1 | - | 8 | 756 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
