siRNA Details
Gene Symbol: TRAPPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021942.5 | NP_068761.4 | 13, 14 | 1561 | ||
NM_199053.2 | NP_951008.1 | 13, 14 | 1561 | |||
XM_017008537.1 | XP_016864026.1 | 13, 14 | 1561 | |||
XM_017008538.1 | XP_016864027.1 | 13, 14 | 1561 | |||
XM_017008539.1 | XP_016864028.1 | 11, 12 | 1212 | |||
GenBank mRNA | AK091146.1 | - | 8, 9 | 785 | ||
AK096345.1 | - | 13, 14 | 1535 | |||
AK315057.1 | - | 8, 9 | 767 | |||
AL136752.1 | - | 5, 6 | 470 | |||
AL833451.1 | - | 12, 13 | 1369 | |||
AL833571.1 | - | 13, 14 | 1540 | |||
BC051724.1 | - | 13, 14 | 1505 | 5265781 | ||
BX538044.1 | - | 13, 14 | 1515 | |||
BX647127.1 | - | 13, 14 | 1532 | |||
BX648499.1 | - | 13, 14 | 1536 | |||
CR533553.1 | - | 5, 6 | 405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
