siRNA Details
Gene Symbol: TRAPPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021942.5 | NP_068761.4 | 22 | 2614 | ||
NM_199053.2 | NP_951008.1 | 22 | 2614 | |||
XM_017008537.1 | XP_016864026.1 | 22 | 2614 | |||
XM_017008538.1 | XP_016864027.1 | 22 | 2614 | |||
XM_017008539.1 | XP_016864028.1 | 20 | 2265 | |||
GenBank mRNA | AK022778.1 | - | 1 | 18 | ||
AK023390.1 | - | 3 | 303 | |||
AK026992.1 | - | 8 | 1006 | |||
AK091146.1 | - | 17 | 1838 | |||
AK096345.1 | - | 22 | 2588 | |||
AK315057.1 | - | 17 | 1820 | |||
AL136752.1 | - | 14 | 1523 | |||
AL833451.1 | - | 21 | 2420 | |||
AL833571.1 | - | 22 | 2593 | |||
BC051724.1 | - | 22 | 2558 | 5265781 | ||
BC139745.1 | - | 10 | 994 | |||
BX538044.1 | - | 22 | 2568 | |||
BX647127.1 | - | 22 | 2585 | |||
BX648499.1 | - | 22 | 2589 | |||
CR533553.1 | - | 14 | 1458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
