siRNA Details
Gene Symbol: TRAPPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021942.5 | NP_068761.4 | 28 | 3315 | ||
NM_199053.2 | NP_951008.1 | 28 | 3315 | |||
XM_017008539.1 | XP_016864028.1 | 26 | 2966 | |||
GenBank mRNA | AK022778.1 | - | 7 | 719 | ||
AK023390.1 | - | 9 | 1004 | |||
AK026992.1 | - | 14 | 1707 | |||
AK090489.1 | - | 3 | 1106 | |||
AK091146.1 | - | 23 | 2539 | |||
AK096345.1 | - | 28 | 3289 | |||
AK315057.1 | - | 23 | 2521 | |||
AL136752.1 | - | 20 | 2224 | |||
AL833451.1 | - | 27 | 3121 | |||
AL833571.1 | - | 29 | 3235 | |||
BC051724.1 | - | 28 | 3259 | 5265781 | ||
BC139745.1 | - | 16 | 1695 | |||
BX538044.1 | - | 28 | 3269 | |||
BX647127.1 | - | 28 | 3286 | |||
BX648499.1 | - | 28 | 3290 | |||
CR533553.1 | - | 20 | 2159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
