siRNA Details
Gene Symbol: TBC1D8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017752.2 | NP_060222.2 | 10 | 1806 | ||
NM_198881.1 | NP_942582.1 | 10 | 1806 | |||
XM_006724669.1 | XP_006724732.1 | 9 | 1656 | |||
XM_006724670.3 | XP_006724733.1 | 10 | 1620 | |||
GenBank mRNA | AB449890.1 | - | 10 | 1641 | ||
AB449891.1 | - | 10 | 1641 | |||
AK091248.1 | - | 1 | 123 | |||
AK123957.1 | - | 10 | 1806 | |||
BC122564.1 | - | 10 | 1805 | 40026331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
