siRNA Details
Gene Symbol: FAM212B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019099.4 | NP_061972.1 | 2 | 271 | ||
NM_198926.2 | NP_945120.1 | 2 | 129 | |||
XM_011541783.2 | XP_011540085.1 | 4 | 367 | |||
GenBank mRNA | AK055667.1 | - | 2 | 247 | ||
AK293989.1 | - | 2 | 156 | |||
AK295012.1 | - | 2 | 210 | |||
AL137198.1 | - | 2 | 178 | |||
AL137199.1 | - | 2 | 129 | |||
BC031558.1 | - | 2 | 226 | 5181834 | ||
DA306915.1 | - | 2 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
