siRNA Details
Gene Symbol: FAM212B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019099.4 | NP_061972.1 | 2 | 1795 | ||
NM_198926.2 | NP_945120.1 | 2 | 1653 | |||
XM_011541783.2 | XP_011540085.1 | 4 | 1891 | |||
GenBank mRNA | AK055667.1 | - | 2 | 1771 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
