siRNA Details
Gene Symbol: HIST1H4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003542.3 | NP_003533.1 | 1 | 142 | ||
GenBank mRNA | BC054014.1 | - | 1 | 198 | 6499185 | |
BC130558.1 | - | 1 | 214 | 40146589 | ||
BC130560.1 | - | 1 | 214 | 40146591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
