siRNA Details
Gene Symbol: APAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181861.1 | NP_863651.1 | 3, 4 | 901 | ||
NM_181868.1 | NP_863658.1 | 3, 4 | 901 | |||
NM_181869.1 | NP_863659.1 | 3, 4 | 901 | |||
XM_017019250.1 | XP_016874739.1 | 3, 4 | 1060 | |||
GenBank mRNA | AB103079.1 | - | 2, 3 | 369 | ||
AF149794.1 | - | 2, 3 | 324 | |||
AJ243003.1 | - | 2, 3 | 324 | |||
AJ243004.1 | - | 2, 3 | 324 | |||
AJ243005.1 | - | 2, 3 | 324 | |||
AJ243006.1 | - | 2, 3 | 324 | |||
AJ243007.1 | - | 2, 3 | 324 | |||
AJ243008.1 | - | 2, 3 | 324 | |||
AJ243009.1 | - | 2, 3 | 324 | |||
AJ243010.1 | - | 2, 3 | 324 | |||
AJ243011.1 | - | 2, 3 | 324 | |||
AJ243048.1 | - | 2, 3 | 324 | |||
AJ243107.1 | - | 2, 3 | 324 | |||
AK307509.1 | - | 3, 4 | 1060 | |||
BC136531.1 | - | 3, 4 | 472 | 9020521 | ||
BC136532.1 | - | 3, 4 | 472 | 9020522 | ||
EF560718.1 | - | 2, 3 | 329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
