siRNA Details
Gene Symbol: APAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160.2 | NP_001151.1 | 6 | 1307 | ||
NM_013229.2 | NP_037361.1 | 6 | 1307 | |||
NM_181861.1 | NP_863651.1 | 6 | 1340 | |||
NM_181868.1 | NP_863658.1 | 6 | 1340 | |||
NM_181869.1 | NP_863659.1 | 6 | 1340 | |||
XM_017019250.1 | XP_016874739.1 | 6 | 1499 | |||
GenBank mRNA | AB007873.2 | - | 5 | 730 | ||
AB103079.1 | - | 5 | 808 | |||
AF013263.1 | - | 6 | 1307 | |||
AF134397.1 | - | 5 | 730 | |||
AF149794.1 | - | 5 | 763 | |||
AJ243003.1 | - | 5 | 763 | |||
AJ243004.1 | - | 5 | 763 | |||
AJ243005.1 | - | 5 | 763 | |||
AJ243006.1 | - | 5 | 763 | |||
AJ243007.1 | - | 5 | 763 | |||
AJ243008.1 | - | 5 | 763 | |||
AJ243009.1 | - | 5 | 763 | |||
AJ243010.1 | - | 5 | 763 | |||
AJ243011.1 | - | 5 | 763 | |||
AJ243048.1 | - | 5 | 763 | |||
AJ243107.1 | - | 5 | 763 | |||
AK307509.1 | - | 6 | 1499 | |||
BC136531.1 | - | 6 | 911 | 9020521 | ||
BC136532.1 | - | 6 | 911 | 9020522 | ||
BC150255.1 | - | 5 | 759 | 9007183 | ||
EF560718.1 | - | 5 | 768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
