siRNA Details
Gene Symbol: HIST1H2AL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003511.2 | NP_003502.1 | 1 | 440 | ||
GenBank mRNA | BC069306.1 | - | 1 | 440 | 7262757 | |
BC112072.1 | - | 1 | 513 | 8327540 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
