siRNA Details
Gene Symbol: SMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002800.2 | NP_001002800.1 | 3 | 607 | ||
NM_005496.3 | NP_005487.3 | 2 | 424 | |||
XM_006713459.3 | XP_006713522.1 | 3 | 377 | |||
XM_011512311.2 | XP_011510613.1 | 3 | 495 | |||
XM_011512312.2 | XP_011510614.1 | 2 | 116 | |||
GenBank mRNA | AB019987.1 | - | 3 | 268 | ||
AK002200.1 | - | 3 | 261 | |||
AK225437.1 | - | 3 | 261 | |||
AK315070.1 | - | 3 | 261 | |||
AL136877.1 | - | 2 | 388 | |||
AL833949.1 | - | 2 | 1315 | |||
BC001557.1 | - | 3 | 243 | |||
BC065259.1 | - | 3 | 439 | |||
BC070161.1 | - | 3 | 261 | |||
BC106033.1 | - | 3 | 279 | |||
BI090399.1 | - | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
