siRNA Details
Gene Symbol: SMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002800.2 | NP_001002800.1 | 6 | 1200 | ||
NM_001288753.1 | NP_001275682.1 | 6 | 1125 | |||
NM_005496.3 | NP_005487.3 | 5 | 1017 | |||
XM_006713459.3 | XP_006713522.1 | 6 | 970 | |||
XM_011512311.2 | XP_011510613.1 | 6 | 1088 | |||
XM_011512312.2 | XP_011510614.1 | 5 | 709 | |||
XM_017005491.1 | XP_016860980.1 | 4 | 535 | |||
GenBank mRNA | AB019987.1 | - | 6 | 861 | ||
AF092564.1 | - | 4 | 491 | |||
AK002200.1 | - | 6 | 854 | |||
AK128143.1 | - | 6 | 779 | |||
AK225437.1 | - | 7 | 854 | |||
AK307508.1 | - | 316 | ||||
AK315070.1 | - | 6 | 854 | |||
AL136877.1 | - | 5 | 981 | |||
AL833949.1 | - | 5 | 1908 | |||
BC001557.1 | - | 6 | 836 | |||
BC065259.1 | - | 6 | 1032 | |||
BC070161.1 | - | 6 | 854 | |||
BC106033.1 | - | 6 | 872 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
