siRNA Details
Gene Symbol: SMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002800.2 | NP_001002800.1 | 14 | 2674 | ||
NM_001288753.1 | NP_001275682.1 | 14 | 2599 | |||
NM_005496.3 | NP_005487.3 | 13 | 2491 | |||
XM_006713459.3 | XP_006713522.1 | 13 | 2210 | |||
XM_011512311.2 | XP_011510613.1 | 14 | 2562 | |||
XM_011512312.2 | XP_011510614.1 | 13 | 2183 | |||
XM_017005491.1 | XP_016860980.1 | 12 | 2405 | |||
GenBank mRNA | AB019987.1 | - | 14 | 2335 | ||
AF092564.1 | - | 12 | 1965 | |||
AK122939.1 | - | 6 | 1447 | |||
AK128143.1 | - | 14 | 2253 | |||
AK307508.1 | - | 1790 | ||||
AK315070.1 | - | 14 | 2330 | |||
AL136877.1 | - | 13 | 2455 | |||
AL833949.1 | - | 13 | 3382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
