siRNA Details
Gene Symbol: TULP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007466.2 | NP_001007467.1 | 1 | 1371 | ||
NM_020245.4 | NP_064630.2 | 1 | 1371 | |||
XM_011535946.1 | XP_011534248.1 | 2 | 2048 | |||
XM_017011063.1 | XP_016866552.1 | 3 | 2197 | |||
XM_017011064.1 | XP_016866553.1 | 4 | 2362 | |||
XM_017011065.1 | XP_016866554.1 | 3 | 2115 | |||
XM_017011066.1 | XP_016866555.1 | 2 | 2005 | |||
XM_017011067.1 | XP_016866556.1 | 2 | 2098 | |||
XM_017011068.1 | XP_016866557.1 | 2 | 2064 | |||
XM_017011069.1 | XP_016866558.1 | 3 | 2200 | |||
XM_017011070.1 | XP_016866559.1 | 2 | 2048 | |||
XM_017011071.1 | XP_016866560.1 | 3 | 2200 | |||
GenBank mRNA | AB037818.1 | - | 1 | 1371 | ||
AF087973.1 | - | 131 | ||||
AF219946.2 | - | 1 | 1371 | |||
AF288480.1 | - | 1 | 1371 | |||
AK304056.1 | - | 1 | 586 | |||
BC152476.1 | - | 1 | 1371 |

More Information
Other Species Targeted

Gene Ontology Categories:
