siRNA Details
Gene Symbol: RHCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB030388.1 | - | 3 | 382 | ||
AB049753.1 | - | 3 | 382 | |||
JF436968.1 | - | 3 | 382 | |||
KF515558.1 | - | 3 | 382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
