siRNA Details
Gene Symbol: RHCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB030388.1 | - | 2 | 205 | ||
AB049753.1 | - | 2 | 205 | |||
AF056965.1 | - | 2 | 205 | |||
AY603478.1 | - | 2 | 221 | |||
BC075081.2 | - | 2 | 226 | 30915384 | ||
BC139905.1 | - | 2 | 333 | 40147407 | ||
DQ266353.1 | - | 2 | 205 | |||
HQ711843.1 | - | 2 | 205 | |||
HQ711844.1 | - | 2 | 225 | |||
HQ711845.1 | - | 2 | 205 | |||
HQ711846.1 | - | 2 | 205 | |||
HQ711847.1 | - | 2 | 225 | |||
JF436968.1 | - | 2 | 205 | |||
JX420291.1 | - | 2 | 205 | |||
JX420292.1 | - | 2 | 205 | |||
JX420295.1 | - | 2 | 205 | |||
JX420296.1 | - | 2 | 205 | |||
JX420300.1 | - | 2 | 205 | |||
JX420302.1 | - | 2 | 205 | |||
KF515558.1 | - | 2 | 205 | |||
KP056313.1 | - | 2 | 205 | |||
X63096.1 | - | 2 | 211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
