siRNA Details
Gene Symbol: CTBP1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033339.1 | 2 | 1583 | |||
GenBank mRNA | AK056133.1 | - | 1 | 2229 | ||
AK125775.1 | - | 3 | 1537 | |||
AL390154.1 | - | 1 | 634 | |||
BC013775.1 | - | 1 | 686 | |||
BC111466.1 | - | 1 | 1031 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
